missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUMB Polyclonal Antibody

NUMB Polyclonal Antibody
Brand: Invitrogen PA587068
This item is not returnable.
View return policy
Description
NUMB Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| NUMB | |
| Polyclonal | |
| Unconjugated | |
| Numb | |
| c14_5527; C14orf41; h-Numb; mnb; m-Nb; m-numb; Nb; NUMB; NUMB endocytic adaptor protein; numb gene homolog; numb gene homolog (Drosophila); numb homolog; numb homolog (Drosophila); NUMB isoform 3; Numb variant 1; Numb variant 2; NUMB, endocytic adaptor protein; protein numb homolog; Protein S171; PTB domain protein; S171 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18222, 29419, 8650 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P49757, Q2LC84, Q9QZS3 | |
| Numb | |
| Recombinant fusion protein of human NUMB. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction