missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NuMA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583112
This item is not returnable.
View return policy
Description
Immunogen sequence: GTPLSITSKL PRTQPDGTSV PGEPASPISQ RLPPKVESLE SLYFTPIPAR SQAPLESSLD SLGDVFLDSG RKTRSARRRT TQIINITMTK KLDVEEPDSA NSSFYSTRSA PASQASLRAT SSTQSLARLG SPDYGN Highest antigen sequence indentity to the following orthologs: Mouse - 90%, Rat - 90%.
NuMA (nuclear mitotic apparatus) is a long coil-coiled protein that plays a role in the interphase and mitosis phases of a cell cycle. In interphase, it has been hypothesized to play a structural role in the nucleoskeleton that may be important in nuclear organization and functions. During mitosis, it is associated with spindle microtubule organization and chromosome positioning.
Specifications
| NuMA | |
| Polyclonal | |
| Unconjugated | |
| NUMA1 | |
| 6720401E04Rik; AA764025; AL022610; AU014979; centrophilin stabilizes mitotic spindle in mitotic cells; NMP22; NMP-22; Nuclear matrix protein-22; Nuclear mitotic apparatus protein; Nuclear mitotic apparatus protein 1; NUMA; NuMA protein; Numa1; SP-H antigen; structural nuclear protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4926 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14980 | |
| NUMA1 | |
| Recombinant protein corresponding to Human NUMA1. Recombinant protein control fragment (Product #RP-92563). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction