missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUFIP2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17752-1-AP-20UL
This item is not returnable.
View return policy
Description
NUFIP2 binds RNA.Specifications
| NUFIP2 | |
| Polyclonal | |
| Unconjugated | |
| NUFIP2 | |
| 182 FIP, 82 FIP, FIP 82, FMRP interacting protein 2, KIAA1321, NUFIP2, PIG1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 57532 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q7Z417 | |
| NUFIP2 | |
| NUFIP2 Fusion Protein Ag12094 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction