missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nuf2 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-99081CL1
This item is not returnable.
View return policy
Description
Nuf2 Polyclonal antibody specifically detects Nuf2 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| Nuf2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| cancer/testis antigen 106, CDCA1hsNuf2, cell division cycle associated 1, Cell division cycle-associated protein 1, CT106, hNuf2, kinetochore protein Nuf2, NUF2, NDC80 kinetochore complex component, homolog (S. cerevisiae), NUF2RhNuf2R | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Nuf2. | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 83540 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction