missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nuf2 Antibody (312), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-43780
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot
Nuf2 Monoclonal specifically detects Nuf2 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Nuf2 | |
| Monoclonal | |
| Unconjugated | |
| PBS with No Preservative | |
| NUF2 | |
| Recombinant protein corresponding to human NUF2 | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 312 | |
| Western Blot 1:500-1:3000, Immunoprecipitation 1:100-1:500 | |
| cancer/testis antigen 106, CDCA1hsNuf2, cell division cycle associated 1, Cell division cycle-associated protein 1, CT106, hNuf2, kinetochore protein Nuf2, NUF2, NDC80 kinetochore complex component, homolog (S. cerevisiae), NUF2RhNuf2R | |
| Mouse | |
| 54 kDa | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 83540 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction