missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£342.00 - £470.00
Specifications
| Antigen | NUDT9 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18437820
|
Novus Biologicals
NBP1-82715-25ul |
25 μL |
£342.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18246607
|
Novus Biologicals
NBP1-82715 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NUDT9 Polyclonal specifically detects NUDT9 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| NUDT9 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| ADP-ribose diphosphatase, ADP-ribose phosphohydrolase, ADP-ribose pyrosphosphatase NUDT9, ADPR-PPase, EC 3.6.1.13, MGC3037, mitochondrial, nucleoside diphosphate linked moiety X-type motif 9, nudix (nucleoside diphosphate linked moiety X)-type motif 9, Nudix motif 9, NUDT10 | |
| NUDT9 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 53343 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:AWMETEAVNYHDETGEIMDNLMLEAGDDAGKVKWVDINDKLKLYASHSQFIKLVAEKRDAHWSEDSEADCHAL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title