missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
NUDT7 Polyclonal specifically detects NUDT7 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | NUDT7 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 2% Sucrose |
| Gene Alias | nudix (nucleoside diphosphate linked moiety X)-type motif 7, peroxisomal coenzyme A diphosphatase NUDT7 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of Human NUDT7 |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?