Learn More
Invitrogen™ NUDT7 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA570718
Product Code. 15889123
  • £440.00 / 100µL
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Goat: 85%; Guinea Pig: 100%; Horse: 92%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
The protein encoded by this gene is a member of the Nudix hydrolase family. Nudix hydrolases eliminate potentially toxic nucleotide metabolites from the cell and regulate the concentrations and availability of many different nucleotide substrates, cofactors, and signaling molecules. Alternatively spliced transcript variants encoding multiple isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NUDT7
Polyclonal
Unconjugated
NUDT7
1300007B24Rik; 2210404C19Rik; coenzyme A diphosphatase; nucleoside diphosphate-linked moiety X motif 7; nudix (nucleoside diphosphate linked moiety X)-type motif 7; nudix hydrolase 7; nudix motif 7; Nudt7; Peroxisomal coenzyme A diphosphatase NUDT7; peroxisomal coenzyme A diphosphatase NUDT7-like
Rabbit
Affinity chromatography
RUO
283927
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
P0C024
NUDT7
synthetic peptide directed towards the N-terminal region of human NUDT7.