missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NUDT6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA537265
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 35 and 17 kDa. Purity is >95% by SDS-PAGE.
NUDT6 (nudix (nucleoside diphosphate linked moiety X)-type motif 6), also known as Antisense basic fibroblast growth factor or GFG-1, is a member of the Nudix hydrolase family of pyrophosphatases. Nudix hydrolases contain a characteristic Nudix domain and are responsible for catalyzing the hydrolysis of nucleoside diphosphate derivatives. The gene encoding NUDT6 is a FGF-2 gene antisense transcript, and NUDT6 is believed to regulate FGF-2 expression. FGF-2 is a mutifunctional heparin-binding growth factor important to angiogenesis, neuroectoderm development and wound healing. NUDT6 is expressed as two isoforms produced by alternative splicing.
Specifications
| NUDT6 | |
| Polyclonal | |
| Unconjugated | |
| NUDT6 | |
| antisense basic fibroblast growth factor; Antisense basic fibroblast growth factor B; anti-sense basic fibroblast growth factor B; ASFGF2; Asfgf2b; FGF2AS; FGF-AS; Gfg; GFG1; GFG-1; Nucleoside diphosphate-linked moiety X motif 6; nudix (nucleoside diphosphate linked moiety X)-type motif 6; nudix hydrolase 6; nudix motif 6; Nudt6; Protein GFG | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11162 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P53370 | |
| NUDT6 | |
| Recombinant full length human NUDT6. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction