missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT6 Monoclonal Antibody (OTI9D12)

NUDT6 Monoclonal Antibody (OTI9D12)
Brand: Invitrogen MA525398
This item is not returnable.
View return policy
Description
NUDT6 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Specifications
| NUDT6 | |
| Monoclonal | |
| 0.75 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P53370 | |
| NUDT6 | |
| Full length human recombinant protein of NUDT6 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI9D12 | |
| Unconjugated | |
| NUDT6 | |
| antisense basic fibroblast growth factor; Antisense basic fibroblast growth factor B; anti-sense basic fibroblast growth factor B; ASFGF2; Asfgf2b; FGF2AS; FGF-AS; Gfg; GFG1; GFG-1; Nucleoside diphosphate-linked moiety X motif 6; nudix (nucleoside diphosphate linked moiety X)-type motif 6; nudix hydrolase 6; nudix motif 6; Nudt6; Protein GFG | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 11162 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction