missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NUDT5 Monoclonal Antibody (739044)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561664
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
Enzyme that can either act as an ADP-sugar pyrophosphatase in absence of diphosphate or catalyze the synthesis of ATP in presence of diphosphate. In absence of diphosphate, hydrolyzes with similar activities various modified nucleoside diphosphates such as ADP-ribose, ADP-mannose, ADP-glucose, 8-oxo-GDP and 8-oxo-dGDP. Can also hydrolyze other nucleotide sugars with low activity. In presence of diphosphate, mediates the synthesis of ATP in the nucleus by catalyzing the conversion of ADP-ribose to ATP and ribose 5-phosphate. Nuclear ATP synthesis takes place when dephosphorylated at Thr-45. Nuclear ATP generation is required for extensive chromatin remodeling events that are energy-consuming. Does not play a role in U8 snoRNA decapping activity. Binds U8 snoRNA.
Specifications
| NUDT5 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9UKK9 | |
| NUDT5 | |
| E. coli-derived recombinant human ADP-Sugar Pyrophosphatase/NUDT5, Glu2-Phe219. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 739044 | |
| Unconjugated | |
| NUDT5 | |
| 8-oxo-dGDP phosphatase; ADP-sugar pyrophosphatase; HSPC115; Nuclear ATP-synthesis protein NUDIX5; Nucleoside diphosphate-linked moiety X motif 5; nudix (nucleoside diphosphate linked moiety X)-type motif 5; nudix hydrolase 5; nudix motif 5; NUDIX5; nudix-type motif 5; NUDT5; YSA1; YSA1H; YSAH1 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 11164 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction