missing translation for 'onlineSavingsMsg'
Learn More

NUDT4 Antibody (OTI1C2) [DyLight 488], Novus Biologicals™

Product Code. 30689328 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30689328 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30689328 Supplier Novus Biologicals Supplier No. NBP345005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NUDT4 Monoclonal antibody specifically detects NUDT4 in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUDT4
Applications Western Blot
Classification Monoclonal
Clone OTI1C2
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias Diadenosine 5'-5'''-P1, diphosphoinositol polyphosphate phosphohydrolase 2, diphosphoinositol polyphosphate phosphohydrolase type 2, DIPP-2, DIPP2alpha, DIPP2beta, DIPP2HDCMB47P, EC 3.6.1.-, EC 3.6.1.52, KIAA0487DKFZp686I1281, Nucleoside diphosphate-linked moiety X motif 4, nudix (nucleoside diphosphate linked moiety X)-type motif 4, Nudix motif 4, P6-hexaphosphate hydrolase 2
Host Species Mouse
Immunogen Full length human recombiant protein of human NUDT4 (NP_061967) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 11163
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.