missing translation for 'onlineSavingsMsg'
Learn More

NUDT4 Antibody (2F2), Novus Biologicals™

Product Code. 18420230 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420230 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18420230 Supplier Novus Biologicals Supplier No. H00011163M08

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NUDT4 Monoclonal antibody specifically detects NUDT4 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUDT4
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2F2
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH12069
Gene Alias Diadenosine 5'-5'''-P1, diphosphoinositol polyphosphate phosphohydrolase 2, diphosphoinositol polyphosphate phosphohydrolase type 2, DIPP-2, DIPP2alpha, DIPP2beta, DIPP2HDCMB47P, EC 3.6.1.-, EC 3.6.1.52, KIAA0487DKFZp686I1281, Nucleoside diphosphate-linked moiety X motif 4, nudix (nucleoside diphosphate linked moiety X)-type motif 4, Nudix motif 4, P6-hexaphosphate hydrolase 2
Host Species Mouse
Immunogen NUDT4 (AAH12069, 1 a.a. ~ 181 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MMKFKPNQTRTYDREGFKKRAACLCFRSEQEDEVLLVSSSRYPDQWIVPGGGMEPEEEPGGAAVREVYEEAGVKGKLGRLLGIFEQNQDRKHRTYVYVLTVTEILEDWEDSVNIGRKREWFKVEDAIKVLQCHKPVHAEYLEKLKLGCSPANGNSTVPSLPDNNALFVTAAQTSGLPSSVR
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 11163
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.