missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT21 Recombinant Rabbit Monoclonal Antibody (11H33L7)
NUDT21 Recombinant Rabbit Monoclonal Antibody (11H33L7)
Brand: Invitrogen 702871
This item is not returnable.
View return policy
Description
NUDT21 Recombinant Monoclonal Antibody for Western Blot
Specifications
| NUDT21 | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| O43809, Q9CQF3 | |
| NUDT21 | |
| Protein corresponding to human NUDT21 [aa21-aa227]. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot | |
| 11H33L7 | |
| Unconjugated | |
| NUDT21 | |
| 25kDa; 3110048P04Rik; 5730530J16Rik; AU014860; AW549947; CFIM25; cleavage and polyadenylation specific factor 5; cleavage and polyadenylation specific factor 5, 25 kD subunit; cleavage and polyadenylation specific factor 5, 25 kDa; cleavage and polyadenylation specificity factor 25 kDa subunit; Cleavage and polyadenylation specificity factor subunit 5; cleavage factor Im complex 25 kDa subunit; CPSF 25 kDa subunit; CPSF25; cpsf5; nucleoside diphosphate-linked moiety X motif 21; nudix (nucleoside diphosphate linked moiety X)-type motif 21; nudix hydrolase 21; Nudix motif 21; Nudt21; pre-mRNA cleavage factor Im (25kD); pre-mRNA cleavage factor Im 25 kDa subunit; Pre-mRNA cleavage factor Im 68 kDa subunit; pre-mRNA cleavage factor Im, 25kD subunit; similar to cleavage and polyadenylation specific factor 5, 25 kDa; zgc:63966 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 11051, 68219 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction