missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NUDT21 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598019
This item is not returnable.
View return policy
Description
NUDT21 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
NUDT21 is one subunit of a cleavage factor required for 3' RNA cleavage and polyadenylation processing. The interaction of the protein with the RNA is one of the earliest steps in the assembly of the 3' end processing complex and facilitates the recruitment of other processing factors.The protein encoded by this gene is one subunit of a cleavage factor required for 3' RNA cleavage and polyadenylation processing. The interaction of the protein with the RNA is one of the earliest steps in the assembly of the 3' end processing complex and facilitates the recruitment of other processing factors. This gene encodes the 25kD subunit of the protein complex, which is composed of four polypeptides.
Specifications
| NUDT21 | |
| Polyclonal | |
| Unconjugated | |
| NUDT21 | |
| 25kDa; 3110048P04Rik; 5730530J16Rik; AU014860; AW549947; CFIM25; cleavage and polyadenylation specific factor 5; cleavage and polyadenylation specific factor 5, 25 kD subunit; cleavage and polyadenylation specific factor 5, 25 kDa; cleavage and polyadenylation specificity factor 25 kDa subunit; Cleavage and polyadenylation specificity factor subunit 5; cleavage factor Im complex 25 kDa subunit; CPSF 25 kDa subunit; CPSF25; cpsf5; nucleoside diphosphate-linked moiety X motif 21; nudix (nucleoside diphosphate linked moiety X)-type motif 21; nudix hydrolase 21; Nudix motif 21; Nudt21; pre-mRNA cleavage factor Im (25kD); pre-mRNA cleavage factor Im 25 kDa subunit; Pre-mRNA cleavage factor Im 68 kDa subunit; pre-mRNA cleavage factor Im, 25kD subunit; similar to cleavage and polyadenylation specific factor 5, 25 kDa; zgc:63966 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 11051 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| O43809 | |
| NUDT21 | |
| Recombinant Human Cleavage and polyadenylation specificity factor subunit 5 protein (2-227AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction