missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | NUDT2 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30227383
|
Novus Biologicals
NBP3-38345-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30229321
|
Novus Biologicals
NBP3-38345-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NUDT2 Polyclonal antibody specifically detects NUDT2 in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| NUDT2 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Apoptosis | |
| PBS (pH 7.3), 50% glycerol | |
| 318 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Ap4A hydrolase, Ap4A hydrolase 1, Ap4Aase, APAH1Diadenosine 5'-5'''-P1, bis(5'-nucleosyl)-tetraphosphatase (asymmetrical), bis(5'-nucleosyl)-tetraphosphatase [asymmetrical], diadenosine 5'-5''-P1, Diadenosine tetraphosphatase, EC 3.6.1.17, MGC10404, Nucleoside diphosphate-linked moiety X motif 2, nudix (nucleoside diphosphate linked moiety X)-type motif 2, Nudix motif 2, P4-tetraphosphate asymmetrical hydrolase, P4-tetraphosphate pyrophosphohydrolase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-147 of human NUDT2 (NP_001152.1).,, Sequence:, MALRACGLIIFRRCLIPKVDNNAIEFLLLQASDGIHHWTPPKGHVEPGEDDLETALRETQEEAGIEAGQLTIIEGFKRELNYVARNKPKTVIYWLAEVKDYDVEIRLSHEHQAYRWLGLEEACQLAQFKEMKAALQEGHQFLCSIEA | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title