missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NUDT19 Recombinant Rabbit Monoclonal Antibody (K01_1U99)
Description
Immunogen sequence: DRSADWLGLF APHHGPPRFG LGPAPFSRTA FPSLPDTDDH KTDNTGTLPE DVAFRICAVR EAFEEAGVLL LRPRTSPPGP APGPGLALEP PPGLASWRDR VRQDPRH Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 58%, Pig - 65%, Cynomolgus monkey - 89%.
NUDT19 is a coenzyme A diphosphatase that mediates the hydrolysis of a wide range of CoA esters, including choloyl-CoA and branched-chain fatty-acyl-CoA esters. At low substrate concentrations medium and long-chain fatty-acyl-CoA esters, NUDT19 are the primary substrates.
Specifications
Specifications
| Antigen | NUDT19 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | K01_1U99 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4 |
| Gene | NUDT19 |
| Gene Accession No. | A8MXV4, Q6AYD9 |
| Gene Alias | CTC-379B2.4; nucleoside diphosphate-linked moiety X motif 19; nucleoside diphosphate-linked moiety X motif 19, mitochondrial; nudix (nucleoside diphosphate linked moiety X)-type motif 19; nudix hydrolase 19; nudix motif 19; NUDT19; RP2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?