missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT19 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55126-1-AP-150UL
This item is not returnable.
View return policy
Description
The antibody is specific to NUDT19.
NUDT19 is a coenzyme A diphosphatase that mediates the hydrolysis of a wide range of CoA esters, including choloyl-CoA and branched-chain fatty-acyl-CoA esters. At low substrate concentrations medium and long-chain fatty-acyl-CoA esters, NUDT19 are the primary substrates.Specifications
| NUDT19 | |
| Polyclonal | |
| Unconjugated | |
| NUDT19 | |
| Nudix motif 19, NUDT19, RP2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 390916 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.59 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| A8MXV4 | |
| NUDT19 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction