missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NUDT18 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555906
This item is not returnable.
View return policy
Description
Immunogen sequence: RLRKNVCYVV LAVFLSEQDE VLLIQEAKRE CRGSWYLPAG RMEPGETIVE ALQREVKEEA GLHCEPETLL SVEERGPSWV RFVFLAR Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.
The protein encoded by this gene is a member of the Nudix hydrolase family. Nudix hydrolases eliminate potentially toxic nucleotide metabolites from the cell and regulate the concentrations and availability of many different nucleotide substrates, cofactors, and signaling molecules. This protein contains a Nudix hydrolase domain and hydrolyzes oxidized forms of guanosine and deoxyguanosine diphosphates.
Specifications
| NUDT18 | |
| Polyclonal | |
| Unconjugated | |
| NUDT18 | |
| 2-hydroxy-dADP phosphatase; 7,8-dihydro-8-oxoguanine phosphatase; 8-oxo-dGDP phosphatase NUDT18; MTH3; MutT homolog 3; mutT human homolog 3; nucleoside diphosphate-linked moiety X motif 18; nudix (nucleoside diphosphate linked moiety X)-type motif 18; nudix hydrolase 18; nudix motif 18; NUDT18 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 79873 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6ZVK8 | |
| NUDT18 | |
| Recombinant protein corresponding to Human NUDT18. Recombinant protein control fragment (Product #RP-93908). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction