missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NUDT14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551323
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total NUDT14 protein.
UDP-glucose (UDPG) acts as the sugar donor in numerous glycosylation reactions, including those involved in the production of glycogen. NUDT14 is a UDPG pyrophosphatase (EC 3.6.1.45) that hydrolyzes UDPG to produce glucose 1-phosphate and UMP (Yagi et al., 2003 [PubMed 12429023]).
Specifications
| NUDT14 | |
| Polyclonal | |
| Unconjugated | |
| NUDT14 | |
| 1110030M18Rik; nucleoside diphosphate-linked moiety X motif 14; nudix (nucleoside diphosphate linked moiety X)-type motif 14; nudix hydrolase 14; nudix motif 14; nudix-type motif 14; NUDT14; UDPG pyrophosphatase; UDP-sugar diphosphatase; UGPP; UGPPase; uridine diphosphate glucose pyrophosphatase; Uridine diphosphate glucose pyrophosphatase NUDT14; uridine diphosphate glucose pyrophosphatase NUDT14; uridine diphosphate glucose pyrophosphatase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 256281 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| O95848 | |
| NUDT14 | |
| Full length fusion protein of human NUDT14. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction