missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NUDT12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA543258
This item is not returnable.
View return policy
Description
Peptide sequence: LALAVSTEIK VDKNEIEDAR WFTREQVLDV LTKGKQQAFF VPPSRAIAHQ Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.
Nucleotides are involved in numerous biochemical reactions and pathways within the cell as substrates, cofactors, and effectors. Nudix hydrolases, such as NUDT12, regulate the concentrations of individual nucleotides and of nucleotide ratios in response to changing circumstances.Nucleotides are involved in numerous biochemical reactions and pathways within the cell as substrates, cofactors, and effectors. Nudix hydrolases, such as NUDT12, regulate the concentrations of individual nucleotides and of nucleotide ratios in response to changing circumstances.
Specifications
| NUDT12 | |
| Polyclonal | |
| Unconjugated | |
| Nudt12 | |
| 0610016O18Rik; DKFZP761I172; MGC137455 protein; nucleoside diphosphate linked moiety X-type motif 12; nucleoside diphosphate-linked moiety X motif 12; nudix (nucleoside diphosphate linked moiety X)-type motif 12; nudix (nucleoside diphosphnudix (nucleoside diphosphate linked moiety X)-type motif 12ate linked moiety X)-type motif 12; nudix hydrolase 12; nudix motif 12; nudix-type motif 12; NUDT12; Peroxisomal NADH pyrophosphatase NUDT12 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 83594 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9BQG2 | |
| Nudt12 | |
| Synthetic peptide directed towards the C-terminal of human NUDT12 (aa 401-450). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction