Learn More
Invitrogen™ NUDT12 Monoclonal Antibody (3F7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA522698
Description
Peptide Sequence: MSSVKRSLKQ EIVTQFHCSA AEGDIAKLTG ILSHSPSLLN ETSENGWTAL MYAARNGHPE IVQFLLEKGC DRSIVNKSRQ TALDIAVFWG YKHIANLLAT AKGGKKPWFL TNEVEECENY FSKTLLDRKS EKRNNSDWLL AKESHPATVF ILFSDLNPLV TLGGNKESFQ QPEVRLCQLN YTDIKDYLAQ PEKITLIFLG VELEIKDKLL NYAGEVPREE EDGLVAWFAL GIDPIAAEEF KQRHENCYFL HPPMPALLQL KEKEAGVVAQ ARSVLAWHSR YKFCPTCGNA TKIEEGGYKR LCLKEDCPSL NGVHNTSYPR VDPVVIMQVI HPDGTKCLLG RQKRFPPGMF TCLAGFIEPG ETIEDAVRRE VEEESGVKVG HVQYVACQPW PMPSSLMIGC LALAVSTEIK VDKNEIEDAR WFTREQVLDV LTKGKQQAFF VPPSRAIAHQ LIKHWIRINP NL.
Nucleotides are involved in numerous biochemical reactions and pathways within the cell as substrates, cofactors, and effectors. Nudix hydrolases, such as NUDT12, regulate the concentrations of individual nucleotides and of nucleotide ratios in response to changing circumstances.Nucleotides are involved in numerous biochemical reactions and pathways within the cell as substrates, cofactors, and effectors. Nudix hydrolases, such as NUDT12, regulate the concentrations of individual nucleotides and of nucleotide ratios in response to changing circumstances.
Specifications
| NUDT12 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9BQG2 | |
| Nudt12 | |
| NUDT12 (AAH41099, 1 a.a. ~ 462 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG3 κ |
| ELISA, Western Blot | |
| 3F7 | |
| Unconjugated | |
| Nudt12 | |
| 0610016O18Rik; DKFZP761I172; MGC137455 protein; nucleoside diphosphate linked moiety X-type motif 12; nucleoside diphosphate-linked moiety X motif 12; nudix (nucleoside diphosphate linked moiety X)-type motif 12; nudix (nucleoside diphosphnudix (nucleoside diphosphate linked moiety X)-type motif 12ate linked moiety X)-type motif 12; nudix hydrolase 12; nudix motif 12; nudix-type motif 12; NUDT12; Peroxisomal NADH pyrophosphatase NUDT12 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 83594 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.