missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT11 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10912-1-AP-20UL
This item is not returnable.
View return policy
Specifications
| NUDT11 | |
| Polyclonal | |
| Unconjugated | |
| NUDT11 | |
| APS1, ASP1, DIPP 3 beta, DIPP3 beta, DIPP3b, DIPP3beta, hAps1, hDIPP3beta, Nudix motif 11, NUDT11 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55190, 58242 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P0C028, Q96G61 | |
| NUDT11 | |
| NUDT11 Fusion Protein Ag1346 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction