missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NUDT11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118129
This item is not returnable.
View return policy
Description
Specificity: Human NUDT11.
Specifications
| NUDT11 | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q96G61 | |
| NUDT11 | |
| E. coli-derived Human NUDT11 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| NUDT11 | |
| APS1; ASP1; Diadenosine 5',5'''-P1,P6-hexaphosphate hydrolase 3-alpha; Diadenosine 5',5'''-P1,P6-hexaphosphate hydrolase 3-beta; diadenosine hexaphosphate hydrolase (AMP-forming); diphosphoinositol polyphosphate phosphohydrolase 3, beta; Diphosphoinositol polyphosphate phosphohydrolase 3-alpha; Diphosphoinositol polyphosphate phosphohydrolase 3-beta; DIPP3; DIPP3 beta; Dipp3a; DIPP-3-alpha; Dipp3b; DIPP3beta; DIPP-3-beta; DIPP3-beta; hAps1; hDIPP3beta; MNCb-1696; Nucleoside diphosphate-linked moiety X motif 10; Nucleoside diphosphate-linked moiety X motif 11; nudix (nucleoside diphosphate linked moiety X)-type motif 11; nudix hydrolase 11; Nudix motif 10; Nudix motif 11; nudix-type motif 11; Nudt10; Nudt11 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 55190 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction