missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: LQCHKPVHAE YLEKLKLGGS PTNGNS Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Cleaves a beta-phosphate from the diphosphate groups in PP-InsP5 (diphosphoinositol pentakisphosphate), suggesting that it may play a role in signal transduction. Also able to catalyze the hydrolysis of dinucleoside oligophosphates, with Ap6A and Ap5A being the preferred substrates. The major reaction products are ADP and p4a from Ap6A and ADP and ATP from Ap5A. Also able to hydrolyze 5-phosphoribose 1-diphosphate.
Specifications
Specifications
| Antigen | NUDT10 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | NUDT10 |
| Gene Accession No. | Q8NFP7 |
| Gene Alias | AI853080; APS2; Diadenosine 5',5'''-P1,P6-hexaphosphate hydrolase 3-alpha; Diadenosine 5',5'''-P1,P6-hexaphosphate hydrolase 3-beta; Diadenosine hexaphosphate hydrolase (AMP-forming); diphosphoinositol polyphosphate phosphohydrolase 3-alpha; Diphosphoinositol polyphosphate phosphohydrolase 3-beta; DIPP3 alpha; Dipp3a; DIPP-3-alpha; DIPP3-alpha; Dipp3b; DIPP-3-beta; hAps2; hDIPP3alpha; MNCb-1696; Nucleoside diphosphate-linked moiety X motif 10; Nucleoside diphosphate-linked moiety X motif 11; nudix (nucleoside diphosphate linked moiety X)-type motif 10; nudix hydrolase 10; nudix motif 10; Nudix motif 11; Nudt10; Nudt11; RGD1564763 |
| Gene Symbols | NUDT10 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
