missing translation for 'onlineSavingsMsg'
Learn More

NUDT11 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18393996 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18393996 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18393996 Supplier Novus Biologicals Supplier No. NBP306300AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NUDT11 Polyclonal specifically detects NUDT11 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUDT11
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias APS1DIPP3B, ASP1, Diadenosine 5'-5'''-P1, diphosphoinositol polyphosphate phosphohydrolase 3, beta, diphosphoinositol polyphosphate phosphohydrolase 3-beta, DIPP3b, DIPP3beta, DIPP-3-beta, DIPP3-beta, EC 3.6.1.-, EC 3.6.1.52, FLJ10628, hDIPP3betahAps1, Nucleoside diphosphate-linked moiety X motif 11, nudix (nucleoside diphosphate linked moiety X)-type motif 11, Nudix motif 11, P6-hexaphosphate hydrolase 3-beta
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human NUDT11 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 55190
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.