missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17508-1-AP-150UL
This item is not returnable.
View return policy
Description
Cleaves a beta-phosphate from the diphosphate groups in PP-InsP5 (diphosphoinositol pentakisphosphate), suggesting that it may play a role in signal transduction. Also able to catalyze the hydrolysis of dinucleoside oligophosphates, with Ap6A and Ap5A being the preferred substrates. The major reaction products are ADP and p4a from Ap6A and ADP and ATP from Ap5A. Also able to hydrolyze 5-phosphoribose 1-diphosphate.Specifications
| NUDT10 | |
| Polyclonal | |
| Unconjugated | |
| NUDT10 | |
| APS2, DIPP 3 alpha, DIPP3 alpha, DIPP3a, hAps2, hDIPP3alpha, Nudix motif 10, NUDT10 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 102954, 170685, 367747 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P0C027, Q8NFP7 | |
| NUDT10 | |
| NUDT10 Fusion Protein Ag11711 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction