missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT10 Antibody (OTI1A6), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73105F
This item is not returnable.
View return policy
Description
NUDT10 Monoclonal antibody specifically detects NUDT10 in Human samples. It is validated for Western Blot
Specifications
| NUDT10 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| OTI1A6 | |
| Western Blot | |
| APS2DIPP3A, Diadenosine 5'-5'''-P1, diphosphoinositol polyphosphate phosphohydrolase 3-alpha, diphosphoinositol polyphosphate phosphohydrolase type 3 alpha, DIPP3a, DIPP-3-alpha, DIPP3-alpha, EC 3.6.1.-, EC 3.6.1.52, hDIPP3alphahAps2, Nucleoside diphosphate-linked moiety X motif 10, nudix (nucleoside diphosphate linked moiety X)-type motif 10, Nudix motif 10, P6-hexaphosphate hydrolase 3-alpha | |
| Full length human recombinant protein of human NUDT10 (NP_694853) produced in E.coli. | |
| 0.1 mL | |
| Signal Transduction | |
| 170685 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction