missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Nudel Monoclonal Antibody (K52041_9G9), Invitrogen™
Mouse Monoclonal Antibody
£416.00 - £734.00
Specifications
| Antigen | Nudel |
|---|---|
| Clone | K52041_9G9 |
| Concentration | 1 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30207689
|
Invitrogen™
MA556001 |
200 μL |
£734.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30194558
|
Invitrogen™
MA555621 |
50 μL |
£416.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: SLKEETAYWK ELSLKYKQSF QEARDELVEF QEGSRELEAE LEAQLVQAEQ RNRDLQADNQ RLKYEVEALK EKLEHQYAQS YKQVSVLEDD LSQTRAIKEQ LHKYVRELEQ ANDDLERAKR ATIVSLEDFE QRLNQAIERN AFLESELDEK ESLLVSVQRL KDEARDLRQE LAVRERQQEV TRKSAPSSPT LDCEKMDSAV QASLSLPATP
Nudel Monoclonal antibody specifically detects Nudel in Human, Mouse, Rat samples. It is validated for Immunoprecipitation, Western BlotSpecifications
| Nudel | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 2600006O07Rik; ATP-sensitive inward rectifier potassium channel 15; DKFZp451M0318; endooligopeptidase A; EOPA; Inward rectifier K(+) channel Kir4.2; Kcnj15; Kir4.2; LIS1-interacting protein NUDEL; LIS1-interacting protein NUDEL; endooligopeptidase A; MITAP1; Mitosin-associated protein 1; mitosin-associated protein MITAP1; mNudel; mNudE-L; NDE1L1; NDE2; Ndel1; nuclear disribution gene E like; nuclear distribution gene E-like homolog 1; nuclear distribution gene E-like homolog 1 (A. nidulans); nuclear distribution protein nudE-like 1; nudE neurodevelopment protein 1 like 1; nudE neurodevelopment protein 1-like 1; nudE nuclear distribution E homolog-like 1; nudE nuclear distribution gene E homolog like 1; nudE nuclear distribution gene E homolog-like 1; Nude2; NUDEL; NUDE-like protein; Potassium channel, inwardly rectifying subfamily J member 15; potassium channel, inwardly rectifying subfamily J, member 15; potassium inwardly-rectifying channel, subfamily J, member 15; Protein mNudE-like; Protein Nudel; unnamed protein product | |
| Recombinant human NDEL1 (immunogen range is 11-220). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Ndel1 |
| K52041_9G9 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| Q78PB6, Q9ERR1, Q9GZM8 | |
| 170845, 81565, 83431 | |
| IgG2b | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title