missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nudel Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-28801
This item is not returnable.
View return policy
Description
Nudel Polyclonal specifically detects Nudel in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| Nudel | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| DKFZp451M0318, EOPA, MITAP1mitosin-associated protein MITAP1, Mitosin-associated protein 1, nuclear distribution protein nudE-like 1, nudE nuclear distribution gene E homolog (A. nidulans)-like 1, nudE nuclear distribution gene E homolog like 1, nudE nuclear distribution gene E homolog like 1 (A. nidulans), NUDELendooligopeptidase A, NUDE-like protein, Protein Nudel | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 81565 | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.1 - 0.3 ug/ml, Peptide ELISA Detection limit 1:8000 | |
| NP_001020750.1 | |
| NDEL1 | |
| Peptide with sequence CRNFAKDQASRKSY corresponding to internal region according to NP_110435.1. | |
| 0.1 mg | |
| Primary | |
| This antibody is expected to recognize both reported isoforms (NP_001020750.1; NP_110435.1). | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction