missing translation for 'onlineSavingsMsg'
Learn More

Nudel Antibody, Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18431255 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18431255 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18431255 Supplier Novus Biologicals Supplier No. NBP176677AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Nudel Polyclonal antibody specifically detects Nudel in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nudel
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias DKFZp451M0318, EOPA, MITAP1mitosin-associated protein MITAP1, Mitosin-associated protein 1, nuclear distribution protein nudE-like 1, nudE nuclear distribution gene E homolog (A. nidulans)-like 1, nudE nuclear distribution gene E homolog like 1, nudE nuclear distribution gene E homolog like 1 (A. nidulans), NUDELendooligopeptidase A, NUDE-like protein, Protein Nudel
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 15 amino acids near the amino terminus of human Nudel. The immunogen is located within the first 50 amino acids of Nudel. Amino Acid Squence: MDGEDIPDFSSLKEE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 81565
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.