missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NUDCD3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554013
This item is not returnable.
View return policy
Description
Immunogen sequence: NPDSYNGAVR ENYTWSQDYT DLEVRVPVPK HVVKGKQVSV ALSSSSIRVA MLEENGERVL MEGKLTHKIN TESSLWSL Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 30%.
NUDCD3 gene ontology annotations related to this gene include cilium assembly; developmental process; protein folding; protein localization to pericentriolar material.
Specifications
| NUDCD3 | |
| Polyclonal | |
| Unconjugated | |
| NUDCD3 | |
| AI427847; BC024322; KIAA1068; KIAA1068 protein; mKIAA1068; nudC domain containing 3; nudC domain-containing protein 3; nudcd3; NudCL; NudC-like protein; Unknown (protein for MGC:137347); zgc:77067 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100125378, 23386 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IVD9 | |
| NUDCD3 | |
| Recombinant protein corresponding to Human NUDCD3. Recombinant protein control fragment (Product #RP-92611). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction