missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
NUDCD2 Polyclonal antibody specifically detects NUDCD2 in Human samples. It is validated for Western Blot, Immunofluorescence
Specifications
Specifications
| Antigen | NUDCD2 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 750 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | DKFZp686E10109, NudC domain containing 2, nudC domain-containing protein 2, NudC-like protein 2 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human NUDCD2 fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?