missing translation for 'onlineSavingsMsg'
Learn More

NUDCD2 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18338326 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18338326 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18338326 Supplier Novus Biologicals Supplier No. NBP306434AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NUDCD2 Polyclonal antibody specifically detects NUDCD2 in Human samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUDCD2
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias DKFZp686E10109, NudC domain containing 2, nudC domain-containing protein 2, NudC-like protein 2
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human NUDCD2 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 134492
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.