missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NUDC Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583001
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Immunogen sequence: MGGEQEEERF DGMLLAMAQQ HEGGVQELVN TFFSFLRRKT DFFIGGEEGM AEKLITQTFS HHNQLAQKTR REKRARQ Highest antigen sequence indentity to the following orthologs: Rat - 97%, Mouse - 97%.
NudC was first identified as a regulator of nuclear movement in the asexual reproductive cycle of the filamentous fungus Aspergillus nidulans. Human NUDC is a nuclear movement protein that associates with dynein.
Tekniske data
| NUDC | |
| Polyclonal | |
| Unconjugated | |
| NUDC | |
| c15; clone 15; H M NPD011; HNUDC; MNUDC; MNUDC protein; NPD011; nuclear distribution C homolog; nuclear distribution C, dynein complex regulator; nuclear distribution gene C homolog; nuclear distribution gene C homolog (Aspergillus); nuclear distribution protein C homolog; Nuclear migration protein nudC; Nudc; nudC nuclear distribution protein; OTTHUMP00000004405; SIG-92; Silg92; silica-induced gene 92 protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10726 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y266 | |
| NUDC | |
| Recombinant protein corresponding to Human NUDC. Recombinant protein control fragment (Product #RP-94045). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion