missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NUDC Recombinant Rabbit Monoclonal Antibody (8A5H4)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA535562
This item is not returnable.
View return policy
Description
Immunogen sequence: SWLIEDGKVV TVHLEKINKM EWWSRLVSSD PEINTKKINP ENSKLSDLDS ETRSMVEKMM YDQRQKSMGL PTSDEQKKQE ILKKFMDQHP EMDFSKAKFN.
NudC was first identified as a regulator of nuclear movement in the asexual reproductive cycle of the filamentous fungus Aspergillus nidulans. Human NUDC is a nuclear movement protein that associates with dynein.
Specifications
| NUDC | |
| Recombinant Monoclonal | |
| 0.31 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O35685, Q63525, Q9Y266 | |
| NUDC | |
| A synthetic peptide corresponding to a sequence within amino acids 232-331 of human NUDC (Q9Y266). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 8A5H4 | |
| Unconjugated | |
| NUDC | |
| c15; clone 15; H M NPD011; HNUDC; MNUDC; MNUDC protein; NPD011; nuclear distribution C homolog; nuclear distribution C, dynein complex regulator; nuclear distribution gene C homolog; nuclear distribution gene C homolog (Aspergillus); nuclear distribution protein C homolog; Nuclear migration protein nudC; Nudc; nudC nuclear distribution protein; OTTHUMP00000004405; SIG-92; Silg92; silica-induced gene 92 protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10726, 18221, 29648 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction