missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDC Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-51971
This item is not returnable.
View return policy
Description
NUDC Polyclonal antibody specifically detects NUDC in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence, ELISA.
Specifications
| NUDC | |
| Polyclonal | |
| Western Blot 0.3 - 1 ug/ml, Immunocytochemistry/Immunofluorescence 10 ug/ml, Peptide ELISA Detection limit 1:32000 | |
| NP_006591.1 | |
| NUDC | |
| Peptide with sequence C-PNLGNGADLPNYR corresponding to internal region according to NP_006591.1. | |
| 0.1 mg | |
| Cell Cycle and Replication | |
| 10726 | |
| Human, Rat | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence, ELISA | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| HNUDC, MNUDC, NPD011, nuclear distribution gene C (A.nidulans) homolog, nuclear distribution gene C homolog (A. nidulans), Nuclear distribution protein C homolog, nuclear migration protein nudC, NudC | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Cross reacts with Human. Expected from sequence similarity: Mouse, Rat, Dog and Cow. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction