missing translation for 'onlineSavingsMsg'
Learn More

Nucleozin, MedChemExpress

Product Code. 16490518
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16490518

Brand: Medchem Express HY50001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Nucleozin, a potent inhibitor of influenza A virus infection, induces the formation of nucleoprotein (NP) aggregates and antagonizes its nuclear accumulation, leading to cessation of viral replication. Nucleozin impedes influenza A virus replication in vitro with a nanomolar EC50.

Nucleozin, a potent inhibitor of influenza A virus infection, induces the formation of nucleoprotein (NP) aggregates and antagonizes its nuclear accumulation, leading to cessation of viral replication. Nucleozin impedes influenza A virus replication in vitro with a nanomolar EC50.

Specifications

Chemical Name or Material Nucleozin
Physical Form Solid
CAS 341001-38-5
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C21H19ClN4O4
For Use With (Application) COVID-19-anti-virus
Solubility Information DMSO : 20 mg/mL (46.85 mM; Need ultrasonic)
SMILES CC1=C(C(C2=CC=CC=C2)=NO1)C(N3CCN(C4=C(C=C(C=C4)[N+]([O-])=O)Cl)CC3)=O
Molecular Weight (g/mol) 426.85
Formula Weight 426.85
Percent Purity 98.81%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.