Learn More
Macherey-Nagel™ NucleoVac Vacuum Regulator
Product Code. 10459561
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Product Code. Quantity Unit Size
10459561 1 Ea. Each
1 options
Product Code. 10459561 Supplier Macherey-Nagel™ Supplier No. 740641
  • ONLINE EXCLUSIVE PRICE
    £157.00
    £124.88 / Pack of 1
    Save £32.12
    20% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Ensures a reliable vacuum source when manually processing DNA columns under vacuum. Macherey-Nagel™ NucleoVac Vacuum Regulator is designed for use with the NucleoVac Vacuum Manifold for fast processing of large sample volumes via parallel processing of columns.
Vacuum filtration columns are used to isolate and purify nucleic acids, including plasmid DNA. A reliable vacuum source is critical for successful manual processing. A vacuum regulator is recommended to control the vacuum pressure levels, which impacts the flow rate of the sample through the column. In addition, a regulator controls the vacuum time to ensure complete filtration of the sample.
The NucleoVac Vacuum Regulator is designed for use with the NucleoVac Vacuum Manifold and with various DNA isolation/purification column systems, including NucleoSpin™, NucleoFast™ and NucleoSnap kits.
Specifications
Product Type NucleoVac Vacuum Regulator
Description For use with NucleoVac Vacuum Manifold and DNA isolation/purification columns (e.g., NucleoSpin™ NucleoFast™ or NucleoSnap™ kits)
For Use With (Application) For controlling of vacuum
Quantity 1 Ea.