missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoVac Mini Adapters
Description
The NucleoVac 24 Vacuum Manifold allows the processing of silica columns for nucleic acid purification by vacuum.
- Silica columns (NucleoSpin™ and NucleoSnap) can be attached to the upper luer slots of the manifold
- Binding and washing steps are carried out by vacuum instead of centrifugation
- Parallel processing of silica columns saves signficant time
- Enables fast processing of large sample volumes
Specifications
Specifications
| Product Type | Leur Adapters |
| Format | NucleoSpin or NucleoSnap Columns |
| Disposable | Single-use |
| For Use With (Equipment) | NucleoVac 24 Vacuum Manifold |
| Quantity | 100 Each |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?