missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoVac 96 Waste Container

Product Code. 15830938
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15830938

Brand: Macherey-Nagel™ 740639

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Waste container for NucleoVac 96 Vacuum Manifold

The NucleoVac 96 Vacuum Manifold is designed for the rapid manual parallel purification of nucleic acids using the 8-well and 96-well NucleoSpin kits and the NucleoFast kits.

  • Based on NucleoSpin silica-membrane technology
  • Allows rapid simultaneous purification of high-quality nucleic acids
  • Time consuming centrifugation and decanting steps are eliminated by use of vacuum
  • Spacer sets provided to accommodate microtiter plates, deep-well blocks (round/square), or microtube racks
  • Waste container is placed inside the vacuum chamber to collect/dispose of waste after drying

Specifications

Product Type Waste Container
Content And Storage 1 Ea., 15°C to 25°C
For Use With (Equipment) NucleoVac 96 Vacuum Manifold
Quantity 1 Ea.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.