Learn More
Macherey-Nagel™ NucleoVac 96 Spacer Set
Product Code. 15830898
Click to view available options
Quantity:
2 x 4 Spacers
Unit Size:
1 set
Product Code. 15830898
Brand: Macherey-Nagel™ 740247
Spacer for processing Microtube Rack, Square‐well Block, Multi 96 Plate / MTP, Deep‐well Block on NucleoVac 96 Vacuum Manifold
The NucleoVac 96 Vacuum Manifold is designed for the rapid manual parallel purification of nucleic acids using the 8-well and 96-well NucleoSpin and NucleoFast kits. Time consuming centrifugation and decanting steps are eliminated by use of vacuum.
  • NucleoVac 96 Vacuum Manifold consists of the manifold base and lid, a spacer set, and a waste container set.
  • Spacer set includes:
    • 2 spacers MTP/MULTI-96 PLATES
    • 2 spacers ROUND-WELL BLOCKS
    • 2 spacers SQUARE-WELL BLOCKS
    • 2 spacers MICROTUBE RACKS
  • For use of NucleoSpin™ 8‐well strips, Starter Set A (column holder and dummy strips) is required
Specifications
Product Type Spacer Set
For Use With (Equipment) NucleoVac 96 Vacuum Manifold
Includes 2 spacers each for: microtiter plates, round-well blocks, square-well blocks, or microtube racks
Quantity 2 x 4 Spacers