missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ VET for viral and bacterial DNA and viral RNA from veterinary samples

Product Code. 17751848
Click to view available options
No. of Reactions:
10
50
250
Unit Size:
10 reactions
250 reactions
50 reactions
This item is not returnable. View return policy

Product Code. 17751848

Brand: Macherey-Nagel™ 740842.50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mini spin kit for the isolation and purification of animal pathogen nucleic acids from veterinary materials.

  • The versatile high-performer for veterinary diagnostic
  • Validated for various sample matrices
  • One size fits all – one standard protocol for all matrices
  • FastTrack protocol for rapid routine testing
  • Isolation of viral and bacterial nucleic acids

Specifications

Capacity 25 μg
Column Type Mini Spin Column
Content And Storage 15°C to 25°C
Description Isolation of viral and bacterial nucleic acids
Format Kit
Scale Mini
Sample Type Animal Tissue, Blood, Cell-free Biological Fluids, Plasma, Serum, Stool, Swab
Elution Volume 100 μL
Enzyme Liquid Proteinase K
Final Product Type Viral and bacterial nucleic acids
For Use With (Equipment) Centrifugation, Vortex Mixer
For Use With (Application) Enzymatic Reactions, NGS, qPCR, qRT−PCR
Includes NucleoSpin columns, buffers, proteinase k
Key Functions Isolation of viral and bacterial nucleic acids
No. of Reactions 50
Nucleic Acid Type Viral and bacterial nucleic acids
Product Type Nucleic acid purification kit
Quantity 50 Preps
Salt Form Chaotropic Salt
Sample Volume Homogenized Sample, 200 μL
Shipping Condition Room temperature
Target Viral and bacterial nucleic acids
Technique Silica membrane technology
Test Time 20 to 40 min.
Yield Recovery 70 to 95%
Starting Material Amount 200 μL
Show More Show Less

For research use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.