missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ Robot-96 Plasmid Plates
Description
NucleoSpin™ Robot-96 Plasmid
- Designed for automated small-scale purification of high-copy plasmid DNA from E. coli
- Allows simultaneous processing of up to 96 samples typically within less than 90 minutes
- For use with common laboratory automation workstations
- Does not require modifications to configuration of compatible robots
- Downstream applications: automated fluorescent sequencing, PCR, or restriction analysis
Specifications
Specifications
| Membrane | Silica |
| Color | Natural |
| Product Type | Binding plates |
| For Use With (Equipment) | NucleoSpin Robot-96 Plasmid kit/core kit; Lab automation equipment |
| Volume (Metric) Working | 75 to 150 μL (Elution Volume) |
| For Use With (Application) | Automated purification of plasmid DNA |
| No. per Pack | 24 Plates |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?