missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ RNA Virus, Midi kit for viral RNA from cell-free fluids

Product Code. 12788612
Click to view available options
Quantity:
1 Each
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 12788612

Brand: Macherey-Nagel™ 740958

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Funnel column kit for time saving parallel isolation of viral RNA / DNA from biological fluids.

The NucleoSpin™ RNA Virus F kit enables the processing of larger volumes. The funnel column allows handling of up to 1 mL serum, plasma and other cell-free solutions with only one loading step, thus reducing cross-contamination. Due to the small diameter of the membrane the elution volume is still between 50 μL to 100 μL (low-salt buffer or water), resulting in a highly concentrated sample directly suitable for any amplification process. The funnel column can be centrifuged in a 50 mL polyethylene tube (included in the kit) and can be closed separately. Elution tubes are fitted directly onto the funnel column. Cross contamination is eliminated because all steps are performed in a closed system. Eight or twelve samples (depending on the centrifuge rotor used) can be processed in 30 minutes. Kit components: NucleoSpin™ funnel columns, elution tubes, 50 mL collecting tubes, RNase-free water, carrier RNA, buffers.
  • Silica membrane technology
  • Typical recovery: >90%
  • Sample material: 1 mL serum, plasma, cell-free biological fluids
  • Fragment size: 100bp to 30kb
  • Elution volume: 50 μL to 100 μL
  • Binding capacity: 30 μg
  • Preparation time: 45 min./2-6 preps
  • Format: funnel columns
  • Support protocol for parallel isolation of viral RNA and DNA (proteinase K required)
  • Applicable for HCV, HBV, HAV, HIV, HSV, HPV, VZV, EBV, parvovirus B19, H5N1 For purification of viral RNA and DNA from serum, plasma, cell-free biological fluids.

Specifications

Format Funnel Column
Final Product Type NucleoSpin™ RNA Virus F Column
For Use With (Application) For Viral RNA from up to 1 mL serum, plasma, cell-free biological fluids
Includes Collection Tubes, collection tubes (50mL), collection tubes (0.5 mL), buffers, carrier RNA
No. of Reactions 25
Product Type NucleoSpin™ RNA Virus F Column
Quantity 1 Each
Sample Size <1 mL
compliance-icons compliance-icons
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.