missing translation for 'onlineSavingsMsg'
Learn More

Macherey Nagel Bioanalysis™ Nucleospin RNA Virus

Product Code. 15136456
This item is not returnable. View return policy

Product Code. 15136456

Brand: Macherey Nagel Bioanalysis™ 740956.240C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mini spin kit for the isolation of viral RNA and DNA from cell-free biological fluids on QIAcube

Macherey Nagel

Specifications

Content And Storage 18°C to 25°C
Format Mini Prep
Sample Type Cell-free Biological Fluids, Plasma, Serum
For Use With (Application) Isolation of Viral Nucleic Acids
No. of Reactions 240 Preps
Size 100 bp−approx. 50 kbp
Target Viral RNA and DNA
Volume (Metric) < 150 L
compliance-icons compliance-icons
Product Identifier
  • NucleoSpin RNA Virus
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash with water thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
Supplemental information
  • MIXTURE LIST-Contains : guanidine hydrochloride, ethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.