missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ RNA Set for NucleoZOL, Mini kit for RNA from NucleoZOL lysates

Product Code. 16381606
Change view
Click to view available options
No. of Reactions:
10
Unit Size:
10 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
16381606 10 10 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
UN 1993 Flammable liquid, n.a.g. (1,4-Dioxan-mixture) Chem class 3 Product Group II
Product Code. 16381606 Supplier Macherey-Nagel™ Supplier No. 740406.10
  • ONLINE EXCLUSIVE PRICE
    £72.75
    £48.70 / 10 reactions
    Save £24.05
    33% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
UN 1993 Flammable liquid, n.a.g. (1,4-Dioxan-mixture) Chem class 3 Product Group II

Mini spin kit for the isolation of RNA from NucleoZOL lysates.

  • Combines NucleoZOL efficiency with proven NucleoSpin™ purity
  • Simple bind-wash-elute procedure
  • Perfect recovery – no loss of total RNA, including miRNA
  • Superior RNA yields due to the efficient lysis using NucleoZOL reagent
  • Save time and benefit from the easy and proven handling
  • Highest quality – even from difficult sample materials
  • NucleoZOL reagent (man. part no. 740404.200) sold separately

Specifications

Format Mini Spin Column Set
Isolation Technology Silica Membrane
Sample Type NucleoZOL Lysate
Efficiency 85-95% recovery
Final Product Type Small RNA (10-200 nt) and large RNA (>200 nt)
For Use With (Equipment) NucleoZOL Reagent (740404.200)
For Use With (Application) RNA purification
Includes 13 mL Binding Buffer MX, 6 mL Wash Buffer RA3 (concentrate), 13 mL RNase-free H2O, 10 NucleoSpin™ RNA Columns (light blue rings – plus Collection Tubes), 10 Collection Tubes (2 mL), 10 Collection Tubes (1.5 mL), and 1 Leaflet
No. of Reactions 10
Sample Size <500 μL NucleoZOL sample
Test Time <1 h
Yield 200 μg binding capacity
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • NucleoSpin RNA Set for NucleoZOL (10)
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contain: 1,4-dioxane

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.