Learn More
Macherey-Nagel™ NucleoSpin™ RNA Plus XS Lysis Buffer
Buffer LB1 for efficient and fast RNA isolation from smallest samples
Brand: Macherey-Nagel™ 740368.30
Additional Details : Weight : 0.03000kg
Description
- RNA isolation from small amounts of cultured cells, human / animal tissue
- RNA from samples stored in RNAlater
- Typical downstream applications: real-time RT-PCR, Northern blotting, primer extension, array technology, RNase protection assays, RNA-sequencing
Specifications
Silica Membrane Technology | |
RNA Purification | |
Buffer |
NucleoSpin™ RNA Plus XS Kits | |
30 mL |
Safety and Handling
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P264-Wash with water thoroughly after handling.
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
For Research Use Only