missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ RNA Plant and Fungi, Mini kit for RNA from plant and fungi

Product Code. 15865244
Change view
Click to view available options
No. of Reactions:
10
50
250
Unit Size:
10 reactions
250 reactions
50 reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
15865244 250 250 reactions
15705543 10 10 reactions
15715543 50 50 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15865244 Supplier Macherey-Nagel™ Supplier No. 740120.250
  • ONLINE EXCLUSIVE PRICE
    £1320.00
    £1087.07 / 250 reactions
    Save £232.93
    17% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Universal kit designed to isolate high-integrity RNA from challenging plant and fungal samples using silica membrane mini spin columns, for use in a variety of downstream applications. Kits with 10, 50 or 250 preps available.

Plant and fungal material is diverse in terms of structure, cell wall composition, biochemical compounds and the types of organs. Common products for RNA isolation often fails to efficiently purify RNA from challenging plant samples.

  • Tailored protocols for diverse starting materials (e.g. seeds, roots, stems, needles)
  • Convenient handling and efficient isolation of high integrity RNA
  • NucleoSpin RNA Plant and Fungi Filters for lysate clearing included
  • Designed for samples rich in secondary metabolites, sugar, starch or other compounds that interfere with nucleic acid purification methods
  • Can also be used for standard samples, such as Arabidopsis leaves
  • Typical downstream applications: real-time RT-PCR, Northern blotting, primer extension, array technology, RNase protection assays

Specifications

Format Mini Spin Column
Isolation Technology Silica Membrane
Elution Volume 50 μL
Sample Type Plant or Fungal Material (Seeds, Roots, Stems, Needles)
Efficiency Binding capacity: 200 μg
Final Product Type High integrity RNA
For Use With (Application) Isolation of RNA from plant and fungal samples
Includes Filters, Columns with Collection Tubes, Collection Tubes (2 mL), Collection Tubes (1.5 mL), buffers, RNase free water
No. of Reactions 250
Purity A260/A280: 1.9 - 2.1
Sample Size < 500 mg
Test Sensitivity RNA integrity number (RIN): 7 - 9
Test Time 25 min / 6 preps (preparation time)
Yield 20 - 70 μg
Show More Show Less
compliance-icons
Product Identifier
  • NucleoSpin RNA Plant and Fungi
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash with water thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
Supplemental information
  • MIXTURE LIST-Contain: guanidine hydrochloride, ethylendinitrilo tetraacetic acid (EDTA)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.