missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ RNA Midi kit for RNA purification
Description
NucleoSpin™ RNA L allows isolation of DNA-free high quality RNA from large amounts of starting material, e.g., cultured cells, tissue, bacteria and yeast. Cells are lysed by incubation in a solution containing large amounts of chaotropic ions. This lysis buffer immediately inactivates RNases and creates appropriate binding conditions which favour adsorption of RNA to the silica membrane. After lysis, homogenisation and reduction of viscosity are achieved by filtration with NucleoSpin™ filter L units. Contaminating DNA bound to the silica membrane is removed by a DNase I solution which is directly applied onto the silica membrane during the preparation. Optimal conditions for the DNase are achieved by washing the silica membrane with a specific desalting buffer before treatment. Salts, metabolites and macromolecular cellular components are removed by simple washing steps with two different buffers. Total RNA is finally eluted with RNase-free water. Kit components: NucleoSpin™ RNA L columns with 15mL collecting tubes, 15mL elution tubes, NucleoSpin™ filters L, buffers, RNase-free DNase I, DNase I reaction buffer, RNase-free water.
- DNase I included for removal of genomic DNA plus NucleoSpin™ filters for homogenisation and reduction of lysate viscosity
- Silica membrane technology
- Yield: 180μg (107 Hela cells), 620μg (4 x 107 Hela cells)
- Sample material: <5 x 107 cultured cells, <1010 bacterial cells, ≤3 x 108 yeast cells, <200mg tissue
- Typical RIN: >9
- Elution volume: 500μL
- Binding capacity: 700μg
- Preparation time: 80 min./4 preps
- Format: midi spin column Total RNA isolation from cultured cells, tissue, bacteria, yeast, samples stored in RNAlater and RNA clean up of reaction mixtures.
Specifications
Specifications
| Bacterial Content | <1010 Cells |
| Format | Midi Spin Column |
| For Use With (Application) | RNA isolation from cultured cells, tissue |
| Includes | NucleoSpin™ RNA Midi Columns with Collection Tubes (15 mL), Collection Tubes (15 mL), NucleoSpin™ Filters Midi, buffers, RNase-free rDNase |
| No. of Reactions | 20 |
| Product Type | NucleoSpin RNA Midi kit |
| Quantity | 20 Pack |
| RNase | Yes |
| Sample Size | <5 x 107 cultured cells, <200 mg human/animal tissue , <1010 bacterial cells, <3 x 108 yeast cells |
| Test Time | 80 min./4 Reaction |
| Show More |
Safety and Handling
Product Identifier
- NucleoSpin RNA Midi (20)
- Danger
- Flammable liquid Category 3
- Acute toxicity Category 4
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1
- LONG-TERM AQUATIC HAZARD Chronic 3
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
- EUH031-Contact with acids liberates toxic gas.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contain: guanidinium thiocyanate, ethanol, rDNase
Product Title
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?